Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: emb-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269902.2 | NP_001256831.1 | 18 - 19 | 4259 | 97 | |
NM_001269903.3 | NP_001256832.1 | 3 - 4 | 608 | 97 | ||
NM_001269904.3 | NP_001256833.1 | 1 - 2 | 164 | 97 | ||
NM_001277003.3 | NP_001263932.1 | 9 - 10 | 1802 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Caenorhabditis elegans Probe spans exons |