Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-79 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001373193.1 | NP_001360177.1 | 3 - 4 | 235 | 148 | |
NM_075333.8 | NP_507734.4 | 7 - 8 | 1029 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Caenorhabditis elegans Probe spans exons |