Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srr-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_075334.3 | NP_507735.1 | 7 - 8 | 1045 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |