Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: phy-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129576.3 | NP_001123048.1 | 1 - 2 | 144 | 58 | |
NM_001129577.3 | NP_001123049.1 | 1 - 2 | 140 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |