Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mrp-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_075411.6 | NP_507812.3 | 13 - 14 | 4512 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Caenorhabditis elegans Probe spans exons |