Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ani-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_075413.4 | NP_507814.1 | 3 - 4 | 773 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Caenorhabditis elegans Probe spans exons |