Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gly-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029045.5 | NP_001024216.1 | 8 - 9 | 1771 | 94 | |
NM_001269959.3 | NP_001256888.1 | 5 - 6 | 1308 | 94 | ||
NM_075449.9 | NP_507850.2 | 5 - 6 | 1310 | 94 | ||
GenBank mRNA | AF031834.1 | - | 5 - 6 | 1323 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Caenorhabditis elegans Probe spans exons |