Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ttx-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029041.3 | NP_001024212.1 | 4 - 5 | 360 | 68 | |
NM_001136442.4 | NP_001129914.1 | 3 - 4 | 247 | 68 | ||
NM_001330887.2 | NP_001317838.1 | 3 - 4 | 322 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons |