Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dcs-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269968.2 | NP_001256897.1 | 1 - 2 | 112 | 75 | |
GenBank mRNA | AY079166.1 | - | 1 - 2 | 111 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |