Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ncx-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047717.6 | NP_001041182.1 | 4 - 5 | 569 | 113 | |
NM_001047718.5 | NP_001041183.1 | 4 - 5 | 571 | 113 | ||
GenBank mRNA | X91803.1 | - | 591 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |