Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mdt-17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269980.3 | NP_001256909.1 | 2 - 3 | 379 | 62 | |
NM_001269981.4 | NP_001256910.1 | 2 - 3 | 379 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Caenorhabditis elegans Probe spans exons |