Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pqn-94 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_075510.5 | NP_507911.1 | 1 - 2 | 109 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Caenorhabditis elegans Probe spans exons |