Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srxa-15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307818.1 | NP_001294747.1 | 2 - 3 | 215 | 126 | |
NM_075588.3 | NP_507989.2 | 2 - 3 | 308 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |