Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dnj-25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270018.2 | NP_001256947.1 | 12 - 13 | 1783 | 68 | |
NM_001270019.3 | NP_001256948.1 | 5 - 6 | 785 | 68 | ||
NM_001270020.4 | NP_001256949.1 | 4 - 5 | 735 | 68 | ||
GenBank mRNA | AF293972.1 | - | 12 - 13 | 1803 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |