Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dnj-25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270018.2 | NP_001256947.1 | 13 - 14 | 1916 | 67 | |
NM_001270019.3 | NP_001256948.1 | 6 - 7 | 918 | 67 | ||
NM_001270020.4 | NP_001256949.1 | 5 - 6 | 868 | 67 | ||
NM_001373209.3 | NP_001360578.1 | 14 | 1929 | 67 | ||
GenBank mRNA | AF293972.1 | - | 13 - 14 | 1936 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |