Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dnj-25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270018.2 | NP_001256947.1 | 5 - 6 | 523 | 63 | |
NM_001373209.3 | NP_001360578.1 | 5 - 6 | 530 | 63 | ||
GenBank mRNA | AF293972.1 | - | 5 - 6 | 543 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |