Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sdf-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_075612.4 | NP_508013.2 | 3 - 4 | 522 | 98 | |
GenBank mRNA | AB108783.1 | - | 3 - 4 | 544 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |