Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: num-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028927.3 | NP_001024098.1 | 5 - 6 | 980 | 81 | |
NM_001136432.5 | NP_001129904.1 | 7 - 8 | 1936 | 81 | ||
NM_001270029.3 | NP_001256958.1 | 4 - 5 | 938 | 81 | ||
NM_075620.6 | NP_508021.1 | 5 - 6 | 1112 | 81 | ||
GenBank mRNA | AF286205.1 | - | 5 - 6 | 1112 | 81 | |
AF286206.1 | - | 5 - 6 | 980 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |