Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028668.4 | NP_001023839.2 | 32 - 33 | 8649 | 122 | |
NM_001028669.4 | NP_001023840.2 | 30 - 31 | 8839 | 122 | ||
NM_001136423.4 | NP_001129895.2 | 31 - 32 | 8559 | 122 | ||
NM_001270032.3 | NP_001256961.1 | 33 - 34 | 8763 | 122 | ||
NM_001270033.2 | NP_001256962.1 | 29 - 30 | 8454 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |