Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pbo-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029068.4 | NP_001024239.1 | 7 - 8 | 1287 | 89 | |
NM_001029069.6 | NP_001024240.1 | 7 - 8 | 1302 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |