Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sor-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_075689.6 | NP_508090.2 | 2 - 3 | 131 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |