Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nrde-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_075691.7 | NP_508092.1 | 2 - 3 | 458 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |