Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ife-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_075693.5 | NP_508094.1 | 3 - 4 | 486 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |