Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: irk-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270047.4 | NP_001256976.2 | 4 - 5 | 404 | 118 | |
NM_001270048.3 | NP_001256977.1 | 4 - 5 | 412 | 118 | ||
NM_001307921.4 | NP_001294850.1 | 4 - 5 | 414 | 118 | ||
NM_001330888.3 | NP_001317907.1 | 5 - 6 | 503 | 118 | ||
NM_001380895.1 | NP_001368610.1 | 5 - 6 | 503 | 118 | ||
NM_001380896.1 | NP_001368348.1 | 5 - 6 | 523 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |