Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mab-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_075773.5 | NP_508174.2 | 3 - 4 | 700 | 65 | |
GenBank mRNA | AY742796.1 | - | 3 - 4 | 651 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Caenorhabditis elegans Probe spans exons |