Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dgk-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001380900.1 | NP_001367482.1 | 9 - 10 | 1724 | 173 | |
NM_001380901.2 | NP_001367480.1 | 12 - 13 | 2335 | 173 | ||
NM_001380902.1 | NP_001367483.1 | 9 - 10 | 1730 | 173 | ||
NM_001380903.1 | NP_001367481.1 | 12 - 13 | 2347 | 173 | ||
NM_001380904.1 | NP_001367484.1 | 10 - 11 | 2099 | 173 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |