Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dop-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_075837.5 | NP_508238.2 | 2 - 3 | 448 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |