Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pdk-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029571.4 | NP_001024742.1 | 1 - 2 | 100 | 91 | |
NM_001029572.4 | NP_001024743.1 | 1 - 2 | 109 | 91 | ||
GenBank mRNA | AF130406.1 | - | 1 - 2 | 100 | 91 | |
AF130407.1 | - | 1 - 2 | 100 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Caenorhabditis elegans Probe spans exons |