Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: str-79 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001373236.2 | NP_001360056.1 | 5 - 6 | 956 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |