Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lsy-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029525.4 | NP_001024696.1 | 3 - 4 | 414 | 92 | |
NM_001029526.4 | NP_001024697.1 | 2 - 3 | 246 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |