Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: crn-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076014.5 | NP_508415.1 | 1 - 2 | 65 | 68 | |
GenBank mRNA | AY303578.1 | - | 1 - 2 | 38 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |