Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: glr-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001350348.3 | NP_001337310.1 | 10 - 11 | 1667 | 75 | |
NM_001373243.1 | NP_001359931.1 | 6 - 7 | 1118 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Caenorhabditis elegans Probe spans exons |