Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fox-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001330894.3 | NP_001317857.1 | 1 | 532 | 97 | |
NM_001330895.5 | NP_001317738.1 | 2906 | 97 | |||
NM_001330896.3 | NP_001317861.1 | 2 - 3 | 265 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |