Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: igcm-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129703.4 | NP_001123175.1 | 5 - 6 | 626 | 83 | |
NM_076078.6 | NP_508479.2 | 5 - 6 | 626 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |