Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cgef-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171641.6 | NP_741736.2 | 7 - 8 | 751 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons |