Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kin-29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076092.6 | NP_508493.1 | 13 - 14 | 2189 | 80 | |
GenBank mRNA | AF403714.1 | - | 13 - 14 | 2121 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |