Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kin-29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076092.6 | NP_508493.1 | 3 - 4 | 371 | 121 | |
GenBank mRNA | AF403714.1 | - | 3 - 4 | 303 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |