Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kin-29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076092.6 | NP_508493.1 | 8 - 9 | 1092 | 64 | |
GenBank mRNA | AF403714.1 | - | 8 - 9 | 1024 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Caenorhabditis elegans Probe spans exons |