Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tra-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383535.1 | 3 - 4 | 183 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Caenorhabditis elegans Probe spans exons |