Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: osm-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076110.7 | NP_508511.1 | 8 - 9 | 1158 | 71 | |
GenBank mRNA | AF314195.1 | - | 8 - 9 | 1143 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |