Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: spc-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270070.3 | NP_001256999.1 | 10 - 11 | 6722 | 75 | |
NM_001270071.5 | NP_001257000.1 | 11 - 12 | 6719 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |