Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: spc-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270070.3 | NP_001256999.1 | 12 - 13 | 7175 | 78 | |
NM_001270071.5 | NP_001257000.1 | 13 - 14 | 7172 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |