Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: glt-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029222.4 | NP_001024393.1 | 5 - 6 | 1040 | 91 | |
NM_001029223.6 | NP_001024394.1 | 4 - 5 | 943 | 91 | ||
GenBank mRNA | D86740.1 | - | 1062 | 91 | ||
D86741.1 | - | 1062 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |