Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: glt-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029222.4 | NP_001024393.1 | 7 - 8 | 1373 | 102 | |
NM_001029223.6 | NP_001024394.1 | 6 - 7 | 1276 | 102 | ||
GenBank mRNA | D86740.1 | - | 1395 | 102 | ||
D86741.1 | - | 1395 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |