Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rgs-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029543.5 | NP_001024714.1 | 2 - 3 | 350 | 83 | |
NM_001373262.3 | NP_001360778.1 | 3 | 426 | 83 | ||
NM_076209.7 | NP_508610.3 | 3 - 4 | 515 | 83 | ||
GenBank mRNA | AB080201.1 | - | 2 - 3 | 381 | 83 | |
AY569308.1 | - | 3 - 4 | 426 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |