Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ckc-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029758.3 | NP_001024929.1 | 4 - 5 | 745 | 93 | |
NM_001029759.5 | NP_001024930.1 | 4 - 5 | 745 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |