Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sax-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029687.6 | NP_001024858.2 | 5 - 6 | 621 | 133 | |
NM_076226.6 | NP_508627.5 | 4 - 5 | 523 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Caenorhabditis elegans Probe spans exons |