Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fkh-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029589.4 | NP_001024760.1 | 3 - 4 | 469 | 84 | |
NM_001029590.6 | NP_001024761.1 | 2 - 3 | 375 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |