Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sup-12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383553.1 | 1 - 2 | 136 | 70 | ||
NM_076273.5 | NP_508674.1 | 1 - 2 | 129 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |