Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: clh-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029714.2 | NP_001024885.1 | 18 - 19 | 2857 | 92 | |
NM_001029715.3 | NP_001024886.1 | 18 - 19 | 2829 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |